Merck
  • Surveillance in ulcerative colitis: is chromoendoscopy-guided endomicroscopy always better than conventional colonoscopy? A randomized trial.
Surveillance in ulcerative colitis: is chromoendoscopy-guided endomicroscopy always better than conventional colonoscopy? A randomized trial.
Inflammatory bowel diseases (2014-09-05)
Paulo Freire, Pedro Figueiredo, Ricardo Cardoso, Maria M Donato, Manuela Ferreira, Sofia Mendes, Mário R Silva, Augusta Cipriano, Ana M Ferreira, Helena Vasconcelos, Francisco Portela, Carlos Sofia
ABSTRACT
Ulcerative colitis (UC) is associated with an increased risk of colorectal cancer. Chromoendoscopy showed superiority to conventional colonoscopy (CC) in surveillance studies including high-risk patients. We aimed to compare chromoendoscopy-guided endomicroscopy (CGE) with CC for intraepithelial neoplasia (IN) detection in patients with longstanding UC without primary sclerosing cholangitis and/or history of IN. One hundred sixty-two patients with longstanding (≥ 8 yr) distal/extensive UC and without primary sclerosing cholangitis and/or history of IN were prospectively randomized to undergo CGE (group A) or CC (group B). Seventeen patients were excluded. In group A (n = 72), circumscribed lesions highlighted by pan-chromoendoscopy were evaluated by endomicroscopy, and targeted biopsy/polypectomy was performed. In group B (n = 73), 4 random biopsies every 10 cm and targeted biopsy/polypectomy of detected lesions were performed. Thirteen IN, all low grade, were detected: 7 IN in group A and 6 in group B (P > 0.05), distributed, respectively, by 6 and 4 patients (P > 0.05). Significantly, more biopsies were performed in group B (4.7 ± 4.9 versus 36.0 ± 6.2, P < 0.001), and the per-biopsy yield of IN was higher in group A (1/48 versus 1/438, P < 0.001). Examination time was 61.5 ± 15.6 minutes in group A and 40.7 ± 8.7 minutes in group B (P < 0.001). The IN detection by endomicroscopy revealed: sensitivity = 85.7%, specificity = 97.9%, positive predictive value = 75.0%, and negative predictive value = 98.9%. CGE does not improve the detection of IN in the endoscopic screening of patients with longstanding UC without primary sclerosing cholangitis and/or history of IN. CGE takes longer than CC, but it decreases the number of biopsies performed and significantly increases the per-biopsy yield of IN. Endomicroscopy is an accurate tool for IN detection.
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology