Merck
  • The monocarboxylate transporter 4 is required for glycolytic reprogramming and inflammatory response in macrophages.
The monocarboxylate transporter 4 is required for glycolytic reprogramming and inflammatory response in macrophages.
The Journal of biological chemistry (2014-11-20)
Zheng Tan, Na Xie, Sami Banerjee, Huachun Cui, Mingui Fu, Victor J Thannickal, Gang Liu
ABSTRACT
There has been fast growing evidence showing that glycolysis plays a critical role in the activation of immune cells. Enhanced glycolysis leads to increased formation of intracellular lactate that is exported to the extracellular environment by monocarboxylate transporter 4 (MCT4). Although the biological activities of extracellular lactate have been well studied, it is less understood how the lactate export is regulated or whether lactate export affects glycolysis during inflammatory activation. In this study, we found that MCT4 is up-regulated by TLR2 and TLR4, but not TLR3 agonists in a variety of macrophages. The increased expression of MCT4 was mediated by MYD88 in a NF-κB-dependent manner. Furthermore, we found that MCT4 is required for macrophage activation upon TLR2 and TLR4 stimulations, as evidenced by attenuated expression of proinflammatory mediators in macrophages with MCT4 knockdown. Mechanistically, we found that MCT4 knockdown leads to enhanced intracellular accumulation of lactate and decreased glycolysis in LPS-treated macrophages. We found that LPS-induced expression of key glycolytic enzymes hexokinase 2 and 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3 is diminished in macrophages with MCT4 knockdown. Our data suggest that MCT4 up-regulation represents a positive feedback mechanism in macrophages to maintain a high glycolytic rate that is essential to a fully activated inflammatory response.
Product Description
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Supelco
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, Ultra pure
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard