Merck
  • Deletion of the major bullous pemphigoid epitope region of collagen XVII induces blistering, autoimmunization, and itching in mice.
Deletion of the major bullous pemphigoid epitope region of collagen XVII induces blistering, autoimmunization, and itching in mice.
The Journal of investigative dermatology (2014-10-14)
Tiina Hurskainen, Nina Kokkonen, Raija Sormunen, Joanna Jackow, Stefanie Löffek, Raija Soininen, Claus-Werner Franzke, Leena Bruckner-Tuderman, Kaisa Tasanen
ABSTRACT
Bullous pemphigoid (BP) is the most common autoimmune subepidermal blistering skin disease with a characteristic of pruritus and blistering. BP patients carry inflammation-triggering autoantibodies against the collagen XVII (ColXVII, also known as BP180) juxtamembraneous extracellular noncollagenous 16A (NC16A) domain involved in ectodomain shedding. Deletion of the corresponding NC14A region in a genetically modified mouse model (ΔNC14A) decreased the amount of ColXVII in skin, but it did not prevent ectodomain shedding. Newborn ΔNC14A mice had no macroscopic phenotypic changes. However, subepidermal microblisters, rudimentary hemidesmosomes, and loose basement membrane zone were observed by microscopy. ΔNC14A mice grow normally, but at around 3 months of age they start to scratch themselves and develop crusted erosions. Furthermore, perilesional eosinophilic infiltrations in the skin, eosinophilia, and elevated serum IgE levels are detected. Despite the removal of the major BP epitope region, ΔNC14A mice developed IgG and IgA autoantibodies with subepidermal reactivity, indicating autoimmunization against a dermo-epidermal junction component. Moreover, IgG autoantibodies recognized a 180-kDa keratinocyte protein, which was sensitive to collagenase digestion. We show here that ΔNC14A mice provide a highly reproducible BP-related mouse model with spontaneous breakage of self-tolerance and development of autoantibodies.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Triton X-100, laboratory grade
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis