Merck
  • PIKfyve, MTMR3 and their product PtdIns5P regulate cancer cell migration and invasion through activation of Rac1.
PIKfyve, MTMR3 and their product PtdIns5P regulate cancer cell migration and invasion through activation of Rac1.
The Biochemical journal (2014-05-21)
Angela Oppelt, Ellen M Haugsten, Tobias Zech, Håvard E Danielsen, Anita Sveen, Viola H Lobert, Rolf I Skotheim, Jørgen Wesche
ABSTRACT
Previously, we have shown that the phosphoinositide metabolizing enzymes PIKfyve (phosphoinositide 5-kinase, FYVE finger containing) and MTMR3 (myotubularin-related protein 3), together with their lipid product PtdIns5P, are important for migration of normal human fibroblasts. As these proteins are a kinase and a phosphatase respectively, and thereby considered druggable, we wanted to test their involvement in cancer cell migration and invasion. First, we showed that PIKfyve and MTMR3 are expressed in most cancer cells. Next, we demonstrated that depletion of PIKfyve or MTMR3 resulted in decreased velocity in three different cancer cell lines by using new software for cell tracking. Inhibition of the enzymatic activity of PIKfyve by the inhibitor YM201636 also led to a strong reduction in cell velocity. Mechanistically, we show that PIKfyve and MTMR3 regulate the activation of the Rho family GTPase Rac1. Further experiments also implicated PtdIns5P in the activation of Rac1. The results suggest a model for the activation of Rac1 in cell migration where PIKfyve and MTMR3 produce PtdIns5P on cellular membranes which may then serve to recruit effectors to activate Rac1. Finally, in an invasion assay, we demonstrate that both PIKfyve and MTMR3 are implicated in invasive behaviour of cancer cells. Thus PIKfyve and MTMR3 could represent novel therapeutic targets in metastatic cancer.
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in NSO cells, suitable for cell culture
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in Baculovirus infected High-5 cells, suitable for cell culture
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution