Merck
  • The effect of hyperglycemia on the pharmacokinetics of valproic acid studied by high-performance liquid chromatography with electrochemical detection.
The effect of hyperglycemia on the pharmacokinetics of valproic acid studied by high-performance liquid chromatography with electrochemical detection.
Journal of pharmaceutical and biomedical analysis (2014-05-13)
Akira Kotani, Tomoko Kotani, Nana Ishii, Hideki Hakamata, Fumiyo Kusu
ABSTRACT
The effects of hyperglycemia on the pharmacokinetics of valproic acid (VPA) were examined by time-concentration profiles of plasma VPA accompanied with blood glucose (BG) changing. In addition, time-concentration profiles of plasma free fatty acids (FFAs) were also obtained to examine the interaction between VPA and FFAs in vivo. For the experiments in vivo, normal rats, given multiple doses of maltose orally, and diabetic rats, which were made to maintain hyperglycemia, were used. Plasma VPA and FFA were determined by high-performance liquid chromatography with electrochemical detection (HPLC-ECD) systems based on the reduction of quinone for the selective determination of acids, respectively. BG was determined by pocket-size glucose meter. The maximum plasma concentrations (Cmax) of VPA in normal rats given multiple doses of maltose orally and in diabetic rats were remarkably decreased in comparison with those in the control rats. From the present study, it was shown that the metabolism of plasma VPA is accelerated under hyperglycemia. Moreover, we also found that VPA was preferentially metabolized in comparison with the plasma FFA in vivo.
Product Description
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Alloxan monohydrate, 98%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Valproic acid sodium salt, 98%
Supelco
Oleic acid, Selectophore, ≥99%
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Stearic acid, analytical standard
Supelco
Oleic acid, analytical standard
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard