Merck
  • Functional characterization of a Plagiochasma appendiculatum flavone synthase I showing flavanone 2-hydroxylase activity.
Functional characterization of a Plagiochasma appendiculatum flavone synthase I showing flavanone 2-hydroxylase activity.
FEBS letters (2014-05-27)
Xiao-Juan Han, Yi-Feng Wu, Shuai Gao, Hai-Na Yu, Rui-Xue Xu, Hong-Xiang Lou, Ai-Xia Cheng
ABSTRACT
FNS I is a 2-oxoglutarate dependent dioxygenase (2-ODD) found mainly in species of the Apiaceae family. Here, an FNS I cDNA sequence was isolated from the liverwort Plagiochasma appendiculatum (Aytoniaceae) and characterized. The recombinant protein exhibited high FNS I activity catalyzing the conversion of naringenin to apigenin and 2-hydroxynaringenin. The critical residue for flavanone-2-hydroxylation activity was Tyr240, as identified from homology modeling and site-directed mutagenesis. The recombinant protein also showed some flavonol synthase activity, as it can convert dihydrokaempferol to kaempferol. When the Leu311 residue was mutated to Phe, the enzyme's capacity to convert dihydrokaempferol to kaempferol was substantially increased. PaFNS I represents a 2-ODD in which a hydrophobic π-stacking interaction between the key residue and the naringenin A-ring determines 2-hydroxyflavanone formation.
Product Description
Supelco
Apigenin, analytical standard
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Naringenin, natural (US), 98%
Sigma-Aldrich
(±)-Naringenin, ≥95%
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Supelco
(±)-Naringenin, analytical standard
Supelco
Kaempferol, analytical standard