Merck
  • CD4 T cells require ICOS-mediated PI3K signaling to increase T-Bet expression in the setting of anti-CTLA-4 therapy.
CD4 T cells require ICOS-mediated PI3K signaling to increase T-Bet expression in the setting of anti-CTLA-4 therapy.
Cancer immunology research (2014-04-30)
Hong Chen, Tihui Fu, Woong-Kyung Suh, Dimitra Tsavachidou, Sijin Wen, Jianjun Gao, Derek Ng Tang, Qiuming He, Jingjing Sun, Padmanee Sharma
ABSTRACT
The transcription factor T-bet controls the Th1 genetic program in T cells for effective antitumor responses. Anti-CTLA-4 immunotherapy elicits dramatic antitumor responses in mice and in human patients; however, factors that regulate T-bet expression during an antitumor response mediated by anti-CTLA-4 remain to be elucidated. We were the first to report that treatment with anti-CTLA-4 led to an increase in the frequency of T cells expressing inducible costimulator (ICOS). In both treated patients and mice, our data revealed that CD4(+)ICOS(hi) T cells can act as effector T cells, which produce the Th1 cytokine IFN-γ. We also showed in a small retrospective analysis that an increased frequency of CD4(+)ICOS(hi) T cells correlated with better clinical outcome and the absence of ICOS or its ligand (ICOSL) in mouse models led to impaired tumor rejection. Here, we show that CD4(+)ICOS(hi) T cells from anti-CTLA-4-treated patients had an increase in signaling via the phospoinositide-3-kinase (PI3K) pathway and an increase in expression of T-bet. An ICOS-specific siRNA transfected into human T cells led to diminished PI3K signaling and T-bet expression. Therefore, we hypothesized that ICOS, and specifically ICOS-mediated PI3K signaling, was required for T-bet expression. We conducted studies in ICOS-deficient and ICOS-YF mice, which have a single amino acid change that abrogates PI3K signaling by ICOS. We found that ICOS-mediated PI3K signaling is required for T-bet expression during an antitumor response elicited by anti-CTLA-4 therapy. Our data provide new insight into the regulation of T-bet expression and suggest that ICOS can be targeted to improve Th1 antitumor responses.
Sigma-Aldrich
MISSION® esiRNA, targeting human CHN1
Sigma-Aldrich
MISSION® esiRNA, targeting human ICOS
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Chn1
Sigma-Aldrich
Propidium iodide solution