Merck
  • Painful nerve injury upregulates thrombospondin-4 expression in dorsal root ganglia.
Painful nerve injury upregulates thrombospondin-4 expression in dorsal root ganglia.
Journal of neuroscience research (2014-10-21)
Bin Pan, Hongwei Yu, John Park, Yanhui Peter Yu, Z David Luo, Quinn H Hogan
ABSTRACT
Thrombospondin-4 (TSP4) belongs to a family of large, oligomeric extracellular matrix glycoproteins that mediate interactions between cells and interactions of cells with underlying matrix components. Recent evidence shows that TSP4 might contribute to the generation of neuropathic pain. However, there has been no systematic examination of TSP4 expression in the dorsal root ganglia (DRG) after injury. This study, therefore, investigates whether TSP4 protein level is changed in DRG after injury following spinal nerve ligation (SNL) and spared nerve injury in rats by performing Western blotting, immunohistochemistry, and immunocytochemistry. After nerve ligation, TSP4 protein level is upregulated in the axotomized somata of the fifth lumbar (L5) DRG. There is substantial additional TSP4 in the nonneuronal compartment of the L5 DRG that does not costain for markers of satellite glia, microglia, or Schwann cells and appears to be in the interstitial space. Evidence of intracellular overexpression of TSP4 persists in neurons dissociated from the L5 DRG after SNL. These findings indicate that, following peripheral nerve injury, TSP4 protein expression is elevated in the cytoplasm of axotomized sensory neurons and in the surrounding interstitial space.
Product Description
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium fluoride 0.5 M solution
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O