Merck
  • Prick testing with chemicals in the diagnosis of occupational contact urticaria and respiratory diseases.
Prick testing with chemicals in the diagnosis of occupational contact urticaria and respiratory diseases.
Contact dermatitis (2014-10-08)
Eva Helaskoski, Hille Suojalehto, Outi Kuuliala, Kristiina Aalto-Korte
ABSTRACT
Little is known about the use of prick tests with chemicals in diagnosing occupational diseases. To evaluate the use of prick tests in the diagnosis of occupational contact urticaria, asthma and rhinitis caused by chemicals (undertaken at the Finnish Institute of Occupational Health). We retrospectively reviewed the patient and test files for the period 1 January 1991 to 31 May 2011. Prick tests were performed with chemical solutions and human serum albumin (HSA)-chemical conjugates. Positive prick test reactions to isocyanate-HSA conjugates were associated with isocyanate-specific IgE in all 20 patients, and 17 patients had a relevant occupational disease. Positive reactions to chloramine-T-HSA conjugates in 10 patients also indicated the presence of specific IgE, although occupational diseases were not always diagnosed. Eleven of 17 patients with positive reactions to persulfate solutions were diagnosed with an occupational disease. Methacrylates, colophonium-related substances, amine hardeners, ethanolamines, glutaraldehyde, glyoxal, pyrocatechol and ammonium thioglycolate did not elicit any relevant prick test reactions. No generalized reactions were detected. Prick tests can be safely used for diagnosing contact urticaria, asthma and rhinitis caused by isocyanates, chloramine-T, persulfates, and chlorhexidine, but the results should be carefully interpreted and related to clinical symptoms and other diagnostic tests.
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
5-Amino-1,3,3-trimethylcyclohexanemethylamine, mixture of cis and trans, ≥99%
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Ethylenediamine, BioXtra
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Ethylenediamine, puriss. p.a., absolute, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Supelco
Ethylenediamine, analytical standard
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥99%
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Hexamethylene diisocyanate, puriss., ≥99.0% (GC)
Sigma-Aldrich
Hexamethylene diisocyanate, purum, ≥98.0% (GC)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)