Merck
  • Scutellariae radix induces apoptosis in chemoresistant SCC-25 human tongue squamous carcinoma cells.
Scutellariae radix induces apoptosis in chemoresistant SCC-25 human tongue squamous carcinoma cells.
The American journal of Chinese medicine (2015-02-03)
Byul-Bora Choi, Jeong Hae Choi, Sang-Rye Park, Ji-Young Kim, Jin-Woo Hong, Gyoo-Cheon Kim
ABSTRACT
Scutellariae radix is one of the most widely used anticancer herbal medicines in several Asian countries, including Korea, Japan, and China. Squamous cell carcinoma (SCC) is one of the most common head and neck carcinomas, which is highly invasive and metastatic, and can potentially develop chemoresistance. Therefore, new effective treatment methods are urgently needed. We determined the effects of Scutellariae radix on SCC-25 cells using the WST-1 assay, F-actin staining, flow cytometry analysis, immunofluorescence staining, and western blot analysis. Scutellariae radix treatment inhibited SCC-25 cell growth in a dose- and time-dependent manner, but it did not inhibit HaCaT (human keratinocyte) cell growth. Changes in cell morphology and disruption of filamentous (F)-actin organization were observed. Scutellariae radix-induced apoptosis as indicated by the translocation of cytochrome c and apoptosis-inducing factor (AIF) into the nucleus and cytosol. Scutellariae radix-induced an increase in cells with sub-G1 DNA content, and increased Bax, cleaved caspase-3, caspase-7, caspase-9, DNA fragmentation factor 45 (DFF 45), and poly(ADP-ribose) polymerase-1 (PARP-1) expression levels. Furthermore, increased expression of phosphorylated mitogen-activated protein kinase (MAPK)-related proteins was detected. The antitumor effect of Scutellariae radix was due to decreased cell proliferation, changes in cell morphology, and the activation of caspase and MAPK pathways. Taken together, the findings of this study highlight the anticancer activity of Scutellariae radix in chemoresistant SCC-25 oral squamous carcinoma cells.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Propidium iodide solution
Etoposide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
5-Fluorouracil, analytical standard
Fluorouracil, European Pharmacopoeia (EP) Reference Standard