Merck
  • Elevated dopamine induces minimal hepatic encephalopathy by activation of astrocytic NADPH oxidase and astrocytic protein tyrosine nitration.
Elevated dopamine induces minimal hepatic encephalopathy by activation of astrocytic NADPH oxidase and astrocytic protein tyrosine nitration.
The international journal of biochemistry & cell biology (2014-09-16)
Saidan Ding, Jianjing Yang, Leping Liu, Yiru Ye, Xuebao Wang, Jiangnan Hu, Bicheng Chen, Qichuan Zhuge
ABSTRACT
We previously demonstrated that dopamine (DA) overload may be a key mechanism behind development of minimal hepatic encephalopathy (MHE) in rats. It has been shown that low-grade cerebral oedema and oxidative stress play important roles in the pathogenesis of MHE. In the current study, DA-triggered oxidative injury in cerebral cortex was studied. An MHE rat model was used. DA was injected intracerebroventricularly (i.c.v.) into rats and added to primary cortical astrocytes (PCAs). Immunoblotting, immunoprecipitation and immunostaining were conducted after DA injection. Cognitive impairment and cerebral edema were observed in MHE rats and rats injected with 10 μg DA. Astrocyte swelling was increased by DA. Astrocytic protein tyrosine nitration (PTN) was induced by DA. DA-induced PTN was insensitive to l-NMMA but was blunted by apocynin, superoxide dismutase, catalase and uric acid. Exposure to DA substantially increased levels of astrocytic NADPH oxidase subunits and induced p47(phox) phosphorylation and reactive oxygen species production but decreased the expression and activity of neuronal-type nitric oxide synthase (nNOS). PTN induced by DA, which was attributed to NADPH oxidase and not to nNOS, may alter astrocyte function and thereby contribute to the precipitation of MHE episodes.
Product Description
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
3-Nitro-L-tyrosine, crystalline
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard