Merck
  • Combination of direct infusion mass spectrometry and gas chromatography mass spectrometry for toxicometabolomic study of red blood cells and serum of mice Mus musculus after mercury exposure.
Combination of direct infusion mass spectrometry and gas chromatography mass spectrometry for toxicometabolomic study of red blood cells and serum of mice Mus musculus after mercury exposure.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-02-11)
M A García-Sevillano, T García-Barrera, F Navarro, N Abril, C Pueyo, J López-Barea, J L Gómez-Ariza
ABSTRACT
Although mercury (Hg) is an important environmental and occupational pollutant, its toxicological effects, especially in serum and red blood cells (RBCs), have been scarcely studied. A toxicometabolomics workflow based on high resolution mass spectrometry approaches has been applied to investigate the toxicological effects of Hg in Mus musculus mice after subcutaneous injection for 10 days, which produced inflammation and vacuolization, steatosis and karyolysis in the hepatic tissue. To this end, direct infusion mass spectrometry (DIMS) of polar and lipophilic extracts from serum and RBCs, using positive and negative mode of acquisition (ESI+/ESI-), and gas chromatography-mass spectrometry were used. A quantitative analysis of reversible oxidized thiols in serum proteins demonstrated a strong oxidative stress induction in the liver of Hg-exposed mice. Endogenous metabolites alterations were identified by partial least squares-discriminant analysis (PLS-DA). Mercury-exposed mice show perturbations in energy metabolism, amino acid metabolism, membrane phospholipid breakdown and oxidative stress-related metabolites in serum along the exposure. This work reports for the first time the effects of Hg-exposure on RBCs metabolic pathways, and reveals disturbances in glycolysis, membrane turnover, glutathione and ascorbate metabolisms.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
Sigma-Aldrich
5-(Iodoacetamido)fluorescein, ≥90% (HPLC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)