Merck
Polaron pair mediated triplet generation in polymer/fullerene blends.
Nature communications (2015-03-05)
Stoichko D Dimitrov, Scot Wheeler, Dorota Niedzialek, Bob C Schroeder, Hendrik Utzat, Jarvist M Frost, Jizhong Yao, Alexander Gillett, Pabitra S Tuladhar, Iain McCulloch, Jenny Nelson, James R Durrant
ABSTRACT
Electron spin is a key consideration for the function of organic semiconductors in light-emitting diodes and solar cells, as well as spintronic applications relying on organic magnetoresistance. A mechanism for triplet excited state generation in such systems is by recombination of electron-hole pairs. However, the exact charge recombination mechanism, whether geminate or nongeminate and whether it involves spin-state mixing is not well understood. In this work, the dynamics of free charge separation competing with recombination to polymer triplet states is studied in two closely related polymer-fullerene blends with differing polymer fluorination and photovoltaic performance. Using time-resolved laser spectroscopic techniques and quantum chemical calculations, we show that lower charge separation in the fluorinated system is associated with the formation of bound electron-hole pairs, which undergo spin-state mixing on the nanosecond timescale and subsequent geminate recombination to triplet excitons. We find that these bound electron-hole pairs can be dissociated by electric fields.
Sigma-Aldrich
1,2-Dichlorobenzene solution, suitable for NMR (reference standard), 5% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
1,2-Dichlorobenzene, suitable for HPLC, 99%
Supelco
1,2-Dichlorobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
1,2-Dichlorobenzene, ReagentPlus®, 99%