Merck
  • 4-Methoxymethyl-2,3,5,6-tetrafluorobenzyl alcohol as a urinary biomarker for monitoring of metofluthrin, a fluorine-containing pyrethroid, in exposed rats.
4-Methoxymethyl-2,3,5,6-tetrafluorobenzyl alcohol as a urinary biomarker for monitoring of metofluthrin, a fluorine-containing pyrethroid, in exposed rats.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals (2015-01-15)
Toshiaki Yoshida
ABSTRACT
A fluorine-containing pyrethroid metofluthrin is widely used recently in mosquito repellents. The urinary excretion kinetics of its metabolites was evaluated in rats to establish an optimal biomarker for monitoring metofluthrin exposure of the general population. After metofluthrin had been administered intraperitoneally to rats, the urinary excretion kinetics of the major metofluthrin metabolites was evaluated by moment analysis. The urinary excretion amounts of 4-methoxymethyl-2,3,5,6-tetrafluorobenzyl alcohol were estimated to be proportional to the absorption amounts over a wide exposure range. Urinary 4-methoxymethyl-2,3,5,6-tetrafluorobenzyl alcohol was considered to be an optimal biomarker for metofluthrin exposure.
Sigma-Aldrich
6-Maleimidohexanoic acid, 90% (GC)
Sigma-Aldrich
1-(Trimethylsilyl)imidazole, ≥98.0%
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
6-Maleimidohexanoic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
1-(Trimethylsilyl)imidazole, 96%
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, ≥95%
Sigma-Aldrich
Chlorotrimethylsilane, Wacker Chemie AG, ≥99.0% (GC)
Supelco
1-(Trimethylsilyl)imidazole, derivatization grade (GC derivatization), LiChropur
Supelco
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, LiChropur, ≥95.0% (GC)
Supelco
Chlorotrimethylsilane, derivatization grade (GC derivatization), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF