Merck
  • Characterization of indoor air quality and resident health in an Arizona senior housing apartment building.
Characterization of indoor air quality and resident health in an Arizona senior housing apartment building.
Journal of the Air & Waste Management Association (1995) (2014-12-17)
Sarah E Frey, Hugo Destaillats, Sebastian Cohn, Sherry Ahrentzen, Matthew P Fraser
ABSTRACT
A survey of key indoor air quality (IAQ) parameters and resident health was carried out in 72 apartments within a single low-income senior housing building in Phoenix, Arizona. Air sampling was carried out simultaneously with a questionnaire on personal habits and general health of residents. Mean PM10 concentrations are 66 +/- 16, 58 +/- 13, and 24 +/- 3 microg/m3 and mean PM2.5 concentrations are 62 +/- 16, 53 +/- 13, and 20 +/- 2 microg/m3 for the living room, kitchen, and outdoor balcony, respectively. Median PM10 concentrations are 17, 18 and 17 microg/m3 and median PM25 concentrations are 13, 14, and 13 microg/m3, respectively. The initial results indicate that increased indoor particle concentrations coincide with residents who report smoking cigarettes. Indoor formaldehyde concentrations revealed median levels of 36.9, 38.8, and 4.3 ppb in the living room, kitchen, and balcony, respectively. Results show that 36% of living room samples and 44% of kitchen samples exceeded the Health Canada REL for chronic exposure to formaldehyde (40 ppb). Associations between occupants' behavior self-reported health conditions, and IAQ are evaluated.
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyde, natural, FG
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)