Merck
  • Enhanced survival of retinal ganglion cells is mediated by Müller glial cell-derived PEDF.
Enhanced survival of retinal ganglion cells is mediated by Müller glial cell-derived PEDF.
Experimental eye research (2014-08-17)
Jan Darius Unterlauft, Thomas Claudepierre, Manuela Schmidt, Katja Müller, Yousef Yafai, Peter Wiedemann, Andreas Reichenbach, Wolfram Eichler
ABSTRACT
The death of retinal ganglion cells (RGC) leads to visual impairment and blindness in ocular neurodegenerative diseases, primarily in glaucoma and diabetic retinopathy; hence, mechanisms that contribute to protecting RGC from ischemia/hypoxia are of great interest. We here address the role of retinal glial (Müller) cells and of pigment-epithelium-derived factor (PEDF), one of the main neuroprotectants released from the glial cells. We show that the hypoxia-induced loss in the viability of cultured purified RGC is due to apoptosis, but that the number of viable RGC increases when co-cultured with Müller glial cells suggesting that glial soluble mediators attenuate the death of RGC. When PEDF was ablated from Müller cells a significantly lower number of RGC survived in RGC-Müller cell co-cultures indicating that PEDF is a major survival factor allowing RGC to escape cell death. We further found that RGC express a PEDF receptor known as patatin-like phospholipase domain-containing protein 2 (PNPLA2) and that PEDF exposure, as well as the presence of Müller cells, leads to an activation of nuclear factor (NF)-κB in RGC. Furthermore, adding an NF-κB inhibitor (SN50) to PEDF-treated RGC cultures reduced the survival of RGC. These findings strongly suggest that NF-κB activation in RGC is critically involved in the pro-survival action of Müller-cell derived PEDF and plays an important role in maintaining neuronal survival.
Sigma-Aldrich
Tricine, BioPerformance Certified, suitable for cell culture, ≥99% (titration)
Sigma-Aldrich
Tricine, ≥99% (titration)
Sigma-Aldrich
Tricine, BioXtra, pH 4.0-6.0 (1 M in H2O), ≥99% (titration)
SAFC
Tricine
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Serpinf1
Sigma-Aldrich
MISSION® esiRNA, targeting human SERPINF1