Merck
  • The diversity of karyotypes and genomes within section Syllinum of the Genus Linum (Linaceae) revealed by molecular cytogenetic markers and RAPD analysis.
The diversity of karyotypes and genomes within section Syllinum of the Genus Linum (Linaceae) revealed by molecular cytogenetic markers and RAPD analysis.
PloS one (2015-04-04)
Nadezhda L Bolsheva, Alexander V Zelenin, Inna V Nosova, Alexandra V Amosova, Tatiana E Samatadze, Olga Yu Yurkevich, Nataliya V Melnikova, Daria A Zelenina, Alexander A Volkov, Olga V Muravenko
ABSTRACT
The wide variation in chromosome number found in species of the genus Linum (2n = 16, 18, 20, 26, 28, 30, 32, 36, 42, 72, 84) indicates that chromosomal mutations have played an important role in the speciation of this taxon. To contribute to a better understanding of the genetic diversity and species relationships in this genus, comparative studies of karyotypes and genomes of species within section Syllinum Griseb. (2n = 26, 28) were carried out. Elongated with 9-aminoacridine chromosomes of 10 species of section Syllinum were investigated by C- and DAPI/С-banding, CMA and Ag-NOR-staining, FISH with probes of rDNA and of telomere repeats. RAPD analysis was also performed. All the chromosome pairs in karyotypes of the studied species were identified. Chromosome DAPI/C-banding patterns of 28-chromosomal species were highly similar. Two of the species differed from the others in chromosomal location of rDNA sites. B chromosomes were revealed in all the 28-chromosomal species. Chromosomes of Linum nodiflorum L. (2n = 26) and the 28-chromosomal species were similar in DAPI/C-banding pattern and localization of several rDNA sites, but they differed in chromosomal size and number. The karyotype of L. nodiflorum was characterized by an intercalary site of telomere repeat, one additional 26S rDNA site and also by the absence of B chromosomes. Structural similarities between different chromosome pairs in karyotypes of the studied species were found indicating their tetraploid origin. RAPD analysis did not distinguish the species except L. nodiflorum. The species of section Syllinum probably originated from a common tetraploid ancestor. The 28-chromosomal species were closely related, but L. nodiflorum diverged significantly from the rest of the species probably due to chromosomal rearrangements occurring during evolution.
Sigma-Aldrich
DAPI, for nucleic acid staining
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Supelco
9-Aminoacridine, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC)
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology