Merck
  • Disulfiram-loaded immediate and extended release vaginal tablets for the localised treatment of cervical cancer.
Disulfiram-loaded immediate and extended release vaginal tablets for the localised treatment of cervical cancer.
The Journal of pharmacy and pharmacology (2014-12-17)
Clara S Baffoe, Nhi Nguyen, Peter Boyd, Weiguang Wang, Mark Morris, Christopher McConville
ABSTRACT
To develop and manufacture both immediate and sustained release vaginal tablets containing the anticancer drug disulfiram, which has the potential to be used as a non-invasive treatment for cervical cancer. Disulfiram-loaded vaginal tablets were manufactured at pilot scale using the direct compression method. These tablets were tested in accordance with the European Pharmacopeia testing of solid dosage form guidelines. They were also tested using a biorelevant dissolution method as well as a dual-chambered release model designed to better mimic the dynamic nature of the vaginal vault. We have developed both immediate and sustained release vaginal tablets, which when manufactured at pilot scale are within the limits set by the European Pharmacopeia for the testing of solid dosage forms. Furthermore, these tablets are capable of releasing disulfiram in vitro using the dual-chambered release model at levels 25,000 times and 35,000 times greater than its IC50 concentration for the HeLa cervical cancer cell line. The successful pilot manufacture and testing of both the immediate and sustained release disulfiram-loaded vaginal tablets warrant further investigation, using an in-vivo model, to assess their potential for use as a non-invasive treatment option for cervical cancer.
Product Description
Stearic acid, European Pharmacopoeia (EP) Reference Standard
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Lactose monohydrate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Tetraethylthiuram disulfide, ≥97.0% (S)
Supelco
Lactose (Monohydrate), Pharmaceutical Secondary Standard; Certified Reference Material
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium stearate, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Disulfiram, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
D-Lactose monohydrate, tested according to Ph. Eur.