Merck
  • T helper cell dysregulation with hepatitis B and rebalance with glucocorticoids.
T helper cell dysregulation with hepatitis B and rebalance with glucocorticoids.
World journal of gastroenterology (2015-01-07)
Zhong-Hua Lu, Xiao-Ping Huang, Wei Sun, Yi-Ling Zhu, Juan-Juan Cui, Wei Chen, Li-Hua Huang, Shou-Gang Kuai, He-Juan Du, Zhao-Xia Ju, Jian-He Gan
ABSTRACT
To investigate T helper 17/regulatory T cell alterations in early severe hepatitis B and the effect of glucocorticoids. The study included 20 patients in the early stage of severe hepatitis B (SHB) and 11 healthy controls. All patients had elevated T helper 17 (Th17) levels, decreased regulatory T (Treg) cell levels, and significant Th17/Treg ratios. After glucocorticoid treatment, 16 patients showed improvement with significant decreases in Th17 levels, increases in Treg, and rebalanced Th17/Treg ratios. The four patients who showed no improvement had increases in both Th17 and Treg levels and an even higher Th17/Treg ratio than before. Glucocorticoid treatment can rectify Th17/Treg dysregulation in patients with SHB.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)