Merck
  • Poly(N-isopropylacrylamide-co-methacrylic acid) pH/thermo-responsive porous hydrogels as self-regulated drug delivery system.
Poly(N-isopropylacrylamide-co-methacrylic acid) pH/thermo-responsive porous hydrogels as self-regulated drug delivery system.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2014-05-23)
Marieta Constantin, Sanda Bucatariu, Valeria Harabagiu, Irina Popescu, Paolo Ascenzi, Gheorghe Fundueanu
ABSTRACT
A dual drug delivery system based on a "biosensor" (pH-sensitive unit) and a delivery component (thermosensitive hydrogel) was developed. The pH/thermosensitive hydrogel is able to restore the thermosensitive characteristics after electrostatic interaction of the pH-sensitive units with selected biologically active compounds that act as triggering agents. The poly(N-isopropylacrylamide-co-methacrylic acid) (poly(NIPAAm-co-MA)) was synthesized as an interesting pH/thermo-responsive copolymer by free radical polymerization method. Due to the presence of carboxylic groups in MA units, the copolymer loses its thermosensitivity at physiological pH and temperature. However, when the negatively-charged carboxylic groups of the pH-sensitive units interact electrostatically with the positively-charged drugs with hydrophobic character propranolol, lidocaine or metoclopramide, taken as model biologically active compounds, the copolymer restores the thermosensitive properties around the physiological pH and temperature. The poly(NIPAAm-co-MA) linear copolymer was converted into pH/thermo-responsive porous microgels using oligomers of NIPAAm above their LCST, as porogens. Accordingly, the swelling/collapsing processes of the microgels occur only after the interaction with the positively-charged hydrophobic drugs. The hydrophobic drug acts as a triggering agent and the pH/temperature sensitive hydrogel turns as a biosensor (pH-sensitive units) and a delivery component (thermosensitive hydrogel).
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Ammonium persulfate, BioUltra, ≥98.0% (RT)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%
Sigma-Aldrich
N-Isopropylacrylamide, ≥99%
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99%
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99.0% (HPLC)
Sigma-Aldrich
N-Isopropylacrylamide, 97%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), purum, ≥98.0% (GC)