Merck
  • Synthesis of [¹¹C]PBR170, a novel imidazopyridine, for imaging the translocator protein with PET.
Synthesis of [¹¹C]PBR170, a novel imidazopyridine, for imaging the translocator protein with PET.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2014-04-02)
Thomas Bourdier, David Henderson, Christopher J R Fookes, Peter Lam, Filomena Mattner, Michael Fulham, Andrew Katsifis
ABSTRACT
The translocator protein (TSPO) ligand 2-(6,8-dichloro-2-(4-ethoxyphenyl)imidazo[1,2-a]pyridin-3-yl)-N-(2-fluoropyridin-3-yl)-N-methylacetamide (PBR170), is a novel imidazopyridineacetamide with high affinity (2.6 nm) and selectivity for the TSPO. The synthesis of [(11)C]PBR170 was accomplished by N-methylation of the corresponding desmethyl precursor with [(11)C]methyl iodide in the presence of sodium hydroxide in dimethylformamide. [(11)C]PBR170 was produced in 30-45% radiochemical yield (decay-corrected, based on [(11)C]methyl iodide) with a radiochemical purity >98% and a specific activity of 90-190 GBq/μmol after 35 min of synthesis time.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material