Merck
  • Stereoselective inhibition of human butyrylcholinesterase by the enantiomers of bambuterol and their intermediates.
Stereoselective inhibition of human butyrylcholinesterase by the enantiomers of bambuterol and their intermediates.
Drug metabolism and disposition: the biological fate of chemicals (2014-12-17)
Marco Pistolozzi, Huaqiao Du, Hong Wei, Wen Tan
ABSTRACT
This work describes the sequential hydrolysis of bambuterol enantiomers and their monocarbamate metabolites (MONO) catalyzed by human butyrylcholinesterase (BChE) as well as the enzyme inhibition resulting from this process. Particular emphasis is given to the contribution given by MONO to the enzyme inhibition because it was not fully characterized in previous works. Bambuterol and MONO enantiomers displayed the same time- and concentration-dependent mechanism of interaction with the enzyme. The hydrolysis kinetics of both bambuterol and MONO was enantioselective, and the (R)-enantiomer of each compound was hydrolyzed fourfold faster than the respective (S)-enantiomer. Even though the enzyme inhibition rates of (R)- and (S)-MONO were much slower than those of their respective bambuterol enantiomers (∼15-fold), both MONO enantiomers showed a significant BChE inhibition when physiologically relevant concentrations of enzyme and inhibitors were used (∼50% of their respective bambuterol enantiomers). The kinetic constants obtained by testing each single compound were used to model the contribution given by MONO to the enzyme inhibition observed for bambuterol. The hydrolysis of MONO enantiomers enhanced the inhibitory power of bambuterol enantiomers of about 27.5% (R) and 12.5% (S) and extended more than 1 hour the duration of inhibition. The data indicate that MONO contribute significantly to the inhibition of BChE occurring in humans upon administration of normal doses of bambuterol. In addition, the hydrolysis of MONO resulted in the rate-limiting step in the conversion of bambuterol in its pharmacologically active metabolite terbutaline; therefore, MONO concentrations should always be monitored during pharmacokinetic studies of bambuterol.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG