Merck
  • The role of extended-release niacin on immune activation and neurocognition in HIV-infected patients treated with antiretroviral therapy - CTN PT006: study protocol for a randomized controlled trial.
The role of extended-release niacin on immune activation and neurocognition in HIV-infected patients treated with antiretroviral therapy - CTN PT006: study protocol for a randomized controlled trial.
Trials (2014-10-09)
Bertrand Lebouché, Mohammad-Ali Jenabian, Joel Singer, Gina M Graziani, Kim Engler, Benoit Trottier, Réjean Thomas, Marie-Josée Brouillette, Jean-Pierre Routy
ABSTRACT
Approximately 30% of HIV-1-infected patients receiving antiretroviral therapy who achieve virologic control have unsatisfactory immune reconstitution, with CD4+ T-cell counts persistently below 350 cells/μL. These patients are at elevated risk for clinical progression to AIDS and non-AIDS events. CD4+ T-cell depletion following infection and persistent immune activation can partially explain this low CD4+ T-cell recovery. Recent data suggest a link between the tryptophan oxidation pathway, immune activation and HIV disease progression based on overstimulation of the tryptophan oxidation pathway by HIV antigens and by interferon-gamma. This overstimulation reduces levels of circulating tryptophan, resulting in inflammation which has been implicated in the development of neurocognitive dysfunction. Niacin (vitamin B3) is able to control the excess tryptophan oxidation, correcting tryptophan depletion, and therefore represents an interesting strategy to improve CD4 recovery.We aim to design a crossover proof-of-concept study to assess supplementation with an extended-release form of niacin (Niaspan FCT™) in combination with antiretroviral therapy, compared to antiretroviral therapy alone, on T-cell immune activation as defined by changes in the percentage of CD8+ CD38+ HLA-DR+ T-cells. This randomized, open-label, interventional crossover study with an immediate versus deferred use of Niaspan FCT for 24 weeks will assess its ability to reduce immune activation and thus increase CD4 recovery in 20 HIV-infected individuals with suboptimal immune responses despite sustained virologic suppression. A substudy evaluating neurocognitive function will also be conducted. This randomized trial will provide an opportunity to evaluate the potential benefit of oral extended-release niacin, a drug that can indirectly increase tryptophan, to reduce immune activation and in turn increase CD4+ T-cell recovery. The study will also allow for the evaluation of the impact of Niaspan FCT on neurocognitive function in HIV-infected individuals with suboptimal immune responses despite sustained virologic suppression. This study was registered with ClinicalTrials.gov on 17 December 2013 (registration number: NCT02018965).
Sigma-Aldrich
Brefeldin A, Ready Made Solution, from Penicillium brefeldianum, 10 mg/mL in DMSO
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, Molecular Biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Nicotinic acid, European Pharmacopoeia (EP) Reference Standard
USP
Niacin, United States Pharmacopeia (USP) Reference Standard
Supelco
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material