Merck
  • Biopharmaceutical evaluation of new slow release tablets obtained by hot tableting of coated pellets with tramadol hydrochloride.
Biopharmaceutical evaluation of new slow release tablets obtained by hot tableting of coated pellets with tramadol hydrochloride.
Acta poloniae pharmaceutica (2014-11-05)
Danuta Szkutnik-Fiedler, Wiesław Sawicki, Monika Balcerkiewicz, Jarosław Mazgalski, Tomasz Grabowski, Edmund Grześkowiak
ABSTRACT
This study was aimed at a biopharmaceutical evaluation of a new oral dosage form of tramadol hydrochloride (TH)--slow release tablets obtained by hot tableting of coated pellets, 100 mg (TP), compared to the conventional slow release tablets, Tramal Retard, 100 mg (TR). Both TP and TR formulations showed a similar release profile of TH (f2 was 71) in in vitro release studies. The in vivo study was a two-treatment, two-period, two-sequence, single-oral dose 100 mg, crossover design using rabbit model with the phases separated by a washout period of 14 days. It was shown that the amount of TH absorbed into the systemic circulation is similar for TP and TR (the 90% confidence intervals for the AUC(0-1), AUC(0-infinity) and ratios were 85-122 and 92-107%, respectively). However, after administration of slow release tablets obtained by hot tableting of coated pellets, a prolonged absorption and elimination processes and a smoother and more extended plasma profile of TH were observed. It can be assumed that the use of a new oral dosage form of TH in patients affects the extension of analgesia after single administration of the drug, with its gradual absorption into the systemic circulation.
Product Description
Tramadol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate
Supelco
Hexane, analytical standard
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%