Merck
  • Preliminary 1 month stability screening of cosmetic multiple emulsions (W/O/W) prepared using cetyl dimethicone copolyol and Polysorbate 80.
Preliminary 1 month stability screening of cosmetic multiple emulsions (W/O/W) prepared using cetyl dimethicone copolyol and Polysorbate 80.
International journal of cosmetic science (2014-10-17)
H Khan, N Akhtar, T Mahmood, A Jameel, S Mohsin
ABSTRACT
The objective of this work was to develop W/O/W emulsions with different concentration of paraffin oil, lipophilic (cetyl dimethicone copolyol) and hydrophilic emulsifiers (polysorbate 80) and to check their stability at different storage conditions. Approximately, 20 formulations (W/O/W) multiple emulsions were prepared, and their stability was checked at different storage conditions for the period of 30 days. Stability of some multiple emulsions ME12, ME13, ME14, ME19 and ME20 was also checked with carbomer as viscosity-enhancing agent. Microscopic analysis for droplet size determination and rheological characterization of most stable multiple emulsions, that is, ME20 were also performed. It was observed that stability of multiple emulsion increases with the addition of gelling agent. Formulation (ME20) with 13.6% paraffin oil, 2.4% cetyl dimethicone copolyol and 0.8% polysorbate 80 was found more stable at 25°C and 40°C for the period of 30 days. Rheological analysis indicated a decrease in viscosity with the passage of time, while droplet size analysis indicated an increase in droplet size with the passage of time. As a conclusion of this work, a stable multiple emulsion with 13.6% paraffin oil, 2.4% cetyl dimethicone copolyol and 0.8% polysorbate 80 can be formulated and can be further studied for any active ingredient for cosmetic purposes.
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Triethylamine, analytical standard