Merck
  • Effect of reversible ligands on oxime-induced reactivation of sarin- and cyclosarin-inhibited human acetylcholinesterase.
Effect of reversible ligands on oxime-induced reactivation of sarin- and cyclosarin-inhibited human acetylcholinesterase.
Toxicology letters (2014-12-20)
Corinna Scheffel, Horst Thiermann, Franz Worek
ABSTRACT
Poisoning by organophosphorus compounds (OP) used as pesticides and nerve agents is due to irreversible inhibition of the enzyme acetylcholinesterase (AChE). Oximes have been widely recognized for their potency to reactivate the inhibited enzyme. The limited efficacy of currently available oximes against a broad spectrum of OP-compounds initiated novel research efforts to improve oxime-based treatment. Hereby, oxime-induced reactivation of OP-inhibited non-human AChE was reported to be accelerated by different AChE-ligands. To investigate this concept with AChE from human source, the inhibitory potency, binding properties and the potential enhancement of oxime-induced reactivation of OP-inhibited AChE by structurally different AChE-ligands was assessed. Several ligands competed with the oxime for the AChE binding-site impairing reactivation of OP-inhibited AChE whereas a markedly accelerated reactivation of sarin-inhibited enzyme by obidoxime was recorded in the presence of edrophonium, galanthamine and donepezil. Enhancement of oxime-induced reactivation with ligands was presumably subject to prevention of re-inhibition by the reaction product phosphonyloxime (POX). In the end, the results of the present study did not confirm that AChE-ligands directly accelerate the reactivation of OP-inhibited AChE by oximes, but indirectly by prevention of re-inhibition by the reaction product POX. This may be due to different experimental conditions and species differences between human and non-human AChE of previous experiments with non-human AChE.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Imipramine hydrochloride, ≥99% (TLC)
Sigma-Aldrich
Imipramine hydrochloride, BioXtra, ≥99% (TLC)
Imipramine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Imipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Pancuronium bromide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
USP
Donepezil hydrochloride, United States Pharmacopeia (USP) Reference Standard
USP
Imipramine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Pancuronium bromide
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Pancuronium bromide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane