Merck
  • Complexation of resveratrol with soy protein and its improvement on oxidative stability of corn oil/water emulsions.
Complexation of resveratrol with soy protein and its improvement on oxidative stability of corn oil/water emulsions.
Food chemistry (2014-05-20)
Zhi-Li Wan, Jin-Mei Wang, Li-Ying Wang, Yang Yuan, Xiao-Quan Yang
ABSTRACT
This work was to evaluate the potential of soy protein isolate (SPI)-resveratrol (RES) complex as an emulsifier to improve the effectiveness of RES as a natural antioxidant in corn oil-in-water emulsions. The physical properties and oxidative stability of emulsions stabilized by the native SPI-RES and heated SPI-RES complexes were evaluated. The water solubility of RES was enhanced by complexation with SPI, which was mainly driven by hydrophobic interactions. Heat treatment favoured the formation of the SPI-RES complex and endowed it with a higher antioxidant activity. Furthermore, the emulsions stabilized by the SPI-RES complexes showed an increased oxidative stability with reduced lipid hydroperoxides and volatile hexanal. This improving effect could be attributed to the targeted accumulation of RES at the oil-water interface accompanied by the adsorption of SPI, as evidenced by the high interfacial RES concentration. These findings show that the soy protein-polyphenol complex exhibited a good potential to act as an efficient emulsifier to improve the oxidative stability of emulsions.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
2,2′-Azobis(2-methylpropionamidine) dihydrochloride, powder or granules, 97%
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%