Merck
  • Efficient production of secreted staphylococcal antigens in a non-lysing and proteolytically reduced Lactococcus lactis strain.
Efficient production of secreted staphylococcal antigens in a non-lysing and proteolytically reduced Lactococcus lactis strain.
Applied microbiology and biotechnology (2014-09-02)
Jolanda Neef, Danny G A M Koedijk, Tjibbe Bosma, Jan Maarten van Dijl, Girbe Buist
ABSTRACT
Cell surface-exposed and secreted proteins are attractive targets for vaccination against pathogenic gram-positive bacteria. To obtain sufficient amounts of such antigens, efficient protein production platforms are needed. In this study, a pipeline for the production and purification of surface-exposed and secreted antigens of the gram-positive bacterial pathogen Staphylococcus aureus is presented. Cytoplasmic or extracellular production of S. aureus antigens was achieved using the Lactococcus lactis strain PA1001, which lacks the major extracellular protease HtrA and the autolysin AcmA to minimize proteolysis and cell lysis, respectively. For most tested S. aureus antigens, secretory production directed by the signal peptide of the major secreted protein Usp45 of L. lactis resulted in higher yields than intracellular production without a signal peptide. Additionally, secretory production of His-tagged antigens allowed their facile one-step purification from the growth medium by metal affinity chromatography. For three of the purified antigens, biological activity was confirmed through enzyme activity assays. We, furthermore, show that the present pipeline can be used to produce staphylococcal antigens with an N-terminal AVI-tag for site-specific labeling with biotin or a C-terminal cell wall-binding domain for cell surface display. We conclude that our L. lactis-based pipeline allows the efficient production of S. aureus antigens and their subsequent purification in one step.
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
1,4-Dithioerythritol, BioUltra, ≥99.0% (RT)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%