Merck
Enzymatic production of three 6-deoxy-aldohexoses from L-rhamnose.
Bioscience, biotechnology, and biochemistry (2014-07-19)
Sirinan Shompoosang, Akihide Yoshihara, Keiko Uechi, Yasuhiko Asada, Kenji Morimoto
ABSTRACT
6-Deoxy-L-glucose, 6-deoxy-L-altrose, and 6-deoxy-L-allose were produced from L-rhamnose with an immobilized enzyme that was partially purified (IE) and an immobilized Escherichia coli recombinant treated with toluene (TT). 6-Deoxy-L-psicose was produced from L-rhamnose by a combination of L-rhamnose isomerase (TT-PsLRhI) and D-tagatose 3-epimerase (TT-PcDTE). The purified 6-deoxy-L-psicose was isomerized to 6-deoxy-L-altrose and 6-deoxy-L-allose with L-arabinose isomerase (TT-EaLAI) and L-ribose isomerase (TT-AcLRI), respectively, and then was epimerized to L-rhamnulose with immobilized D-tagatose 3-epimerase (IE-PcDTE). Following purification, L-rhamnulose was converted to 6-deoxy-L-glucose with D-arabinose isomerase (TT-BpDAI). The equilibrium ratios of 6-deoxy-L-psicose:6-deoxy-L-altrose, 6-deoxy-L-psicose:6-deoxy-L-allose, and L-rhamnulose:6-deoxy-L-glucose were 60:40, 40:60, and 27:73, respectively. The production yields of 6-deoxy-L-glucose, 6-deoxy-L-altrose, and 6-deoxy-L-allose from L-rhamnose were 5.4, 14.6, and 25.1%, respectively. These results indicate that the aldose isomerases used in this study acted on 6-deoxy aldohexoses.
Product Description
Millipore
D-(−)-Arabinose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-(−)-Arabinose, ≥98% (GC)
Sigma-Aldrich
L-(+)-Arabinose, 99%
Sigma-Aldrich
L-(+)-Arabinose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
L-(+)-Arabinose, 98%
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
L-(+)-Ribose, ≥98% (GC)
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG