Merck
  • Preparation and preliminary evaluation of 63Zn-zinc citrate as a novel PET imaging biomarker for zinc.
Preparation and preliminary evaluation of 63Zn-zinc citrate as a novel PET imaging biomarker for zinc.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2014-07-23)
Timothy R DeGrado, Mukesh K Pandey, John F Byrne, Hendrik P Engelbrecht, Huailei Jiang, Alan B Packard, Kevin A Thomas, Mark S Jacobson, Geoffrey L Curran, Val J Lowe
ABSTRACT
Abnormalities of zinc homeostasis are indicated in many human diseases. A noninvasive imaging method for monitoring zinc in the body would be useful to understand zinc dynamics in health and disease. To provide a PET imaging agent for zinc, we have investigated production of (63)Zn (half-life, 38.5 min) via the (63)Cu(p,n)(63)Zn reaction using isotopically enriched solutions of (63)Cu-copper nitrate. A solution target was used for rapid isolation of the (63)Zn radioisotope from the parent (63)Cu ions. Initial biologic evaluation was performed by biodistribution and PET imaging in normal mice. To produce (63)Zn, solutions of (63)Cu-copper nitrate in dilute nitric acid were irradiated by 14-MeV protons in a low-energy cyclotron. An automated module was used to purify (63)Zn from (63)Cu in the target solution. The (63)Cu-(63)Zn mixture was trapped on a cation-exchange resin and rinsed with water, and the (63)Zn was eluted using 0.05 N HCl in 90% acetone. The resulting solution was neutralized with NaHCO3, and the (63)Zn was then trapped on a carboxymethyl cartridge, washed with water, and eluted with isotonic 4% sodium citrate. Standard quality control tests were performed on the product according to current good manufacturing practice, including radionuclidic identity and purity, and measurement of nonradioactive Zn(+2), Cu(+2), Fe(+3), and Ni(+2) by ion-chromatography high-performance liquid chromatography. Biodistribution and PET imaging studies were performed in B6.SJL mice after intravenous administration of (63)Zn-zinc citrate. (63)Cu target material was recycled by eluting the initial resin with 4N HNO3. Yields of 1.07 ± 0.22 GBq (uncorrected at 30-36 min after end of bombardment) of (63)Zn-zinc citrate were obtained with a 1.23 M (63)Cu-copper nitrate solution. Radionuclidic purity was greater than 99.9%, with copper content lower than 3 μg/batch. Specific activities were 41.2 ± 18.1 MBq/μg (uncorrected) for the (63)Zn product. PET and biodistribution studies in mice at 60 min showed expected high uptake in the pancreas (standard uptake value, 8.8 ± 3.2), liver (6.0 ± 1.9), upper intestine (4.7 ± 2.1), and kidney (4.2 ± 1.3). A practical and current good manufacturing practice-compliant preparation of radionuclidically pure (63)Zn-zinc citrate has been developed that will enable PET imaging studies in animal and human studies. (63)Zn-zinc citrate showed the expected biodistribution in mice.
Product Description
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, ACS reagent, 70%