Merck
  • Phenylbutazone and ketoprofen binding to serum albumin. Fluorescence study.
Phenylbutazone and ketoprofen binding to serum albumin. Fluorescence study.
Pharmacological reports : PR (2014-08-26)
Małgorzata Maciążek-Jurczyk
ABSTRACT
A combination of phenylbutazone (PBZ) and ketoprofen (KP) is popular in therapy of rheumatoid arthritis (RA) but could be unsafe due to the uncontrolled growth of toxicity. Quenching fluorescence of serum albumin in the presence of the both drugs has been characterized by dynamic KQ [M(-1)], static V [M(-1)] quenching constants and also association constants Ka [M(-1)]. The quenching of tryptophanyl residues fluorescence by the KP and PBZ indicates the capability of these drugs to accept the energy from Trp-214 and Trp-135. Strong displacement of KP and PBZ bound to albumin cause by the binding of the second drug to SA close to Trp-214 (subdomain IIA) has been obtained. The displacement was also confirmed on the basis of quenching and association constants. The conclusion, that both PBZ and KP form a binding site in the same subdomains (IIA or/and IB), points to the necessity of using a monitoring therapy owning to the possible increase of the uncontrolled toxic effects.
Product Description
Ketoprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Ketoprofen, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ketoprofen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Supelco
Ketoprofen, VETRANAL®, analytical standard
Sigma-Aldrich
Ketoprofen, meets USP testing specifications
Phenylbutazone, European Pharmacopoeia (EP) Reference Standard
Supelco
Phenylbutazone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Phenylbutazone