Merck
The removal of Triton X-100 by dialysis is feasible!
Analytical and bioanalytical chemistry (2014-11-27)
Stefan Opitz, Franziska Hannika, Thomas Krüger, Heidrun Rhode
ABSTRACT
Triton X-100 has been widely used in many analytical and preparative protocols for a long time. Nevertheless, mass spectrometry, chromatographic separation, and spectrophotometric readout may be considerably hampered by this detergent due to signal suppression, complex formation, and high blank values, respectively. Additionally, Triton X-100 is not safe to remove prior to analytics. Here, microdialysis is introduced as a parallelizable, high-throughput method to clean samples from Triton X-100 with high efficacy and precision. To achieve this, we exploit the potential to considerably increase the critical micellar concentration of Triton X-100 by alteration of matrix properties. To that end, addition of several chaotropic compounds and organic solvents has been shown to increase the critical micellar concentration as well as the removal rate of the detergent. For application, matrix additives can be selected for analyte stability requirements out of a variety of compounds. Conveniently, all these additives are removable subsequently using the same microdialysis tool for downstream analytics requirements. Applicability and protocols are shown with proteomic sample preparation of purified proteins and complex protein mixtures prior to matrix-assisted laser desorption ionization (MALDI) mass spectrometry.
Product Description
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Supelco
2-Propanol, analytical standard
Supelco
Trifluoroacetic acid, analytical standard
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets