Merck
  • Sphingobium endophyticus sp. nov., isolated from the root of Hylomecon japonica.
Sphingobium endophyticus sp. nov., isolated from the root of Hylomecon japonica.
Antonie van Leeuwenhoek (2015-01-28)
Lingfang Zhu, Kaiyun Xin, Chaoqiong Chen, Changfu Li, Meiru Si, Liang Zhao, Xu Shi, Lei Zhang, Xihui Shen
ABSTRACT
A yellow-pigmented bacterium, designated strain GZGR-4(T), was isolated from the root of Hylomecon japonica (Thunb.) Prantl et Kündig collected from Taibai Mountain in Shaanxi Province, north-west China. Cells of strain GZGR-4(T) were Gram-negative, rod-shaped, strictly aerobic, non-endospore-forming and non-motile. Phylogenetic analyses based on 16S rRNA gene sequences showed that strain GZGR-4(T) is a member of the genus Sphingobium, exhibiting the highest sequence similarity to Sphingobium aromaticiconvertens DSM 12677(T) (97.3 %). 16S rRNA gene sequence similarities between strain GZGR-4(T) and the type strains of other Sphingobium species with validly published names ranged from 93.4-96.5 %. The predominant respiratory quinone of strain GZGR-4(T) was ubiquinone-10 (Q-10) and the major cellular fatty acids were summed feature 8 (comprising C18:1 ω7c and/or C18:1 ω6c), summed feature 3 (comprising C16:1 ω7c and/or C16:1 ω6c), C16:0 and C14:0 2-OH. Spermidine was the major polyamine. The polar lipid profile consisted of phosphatidylglycerol, diphosphatidylglycerol, phosphatidylethanolamine, phosphatidylcholine, sphingoglycolipid, one unidentified phosphoglycolipid, one unidentified phospholipid, one unidentified aminolipid and one unidentified lipid. The DNA G+C content was 63.6 mol%. DNA-DNA relatedness for strain GZGR-4(T) with respect to its closest phylogenetic relative S. aromaticiconvertens DSM 12677(T) was 22.6 ± 5.3 %. On the basis of the polyphasic taxonomic data presented, strain GZGR-4(T) is considered to represent a novel species of the genus Sphingobium, for which the name Sphingobium endophyticus sp. nov. is proposed. The type strain is GZGR-4(T) (=CCTCC AB 2013305(T) = KCTC 32447(T)).
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
L-Tyrosine, FG
SAFC
L-Tyrosine
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
USP
L-Tyrosine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%