Merck
  • Enzymatic conversion from pyridoxal to pyridoxine caused by microorganisms within tobacco phyllosphere.
Enzymatic conversion from pyridoxal to pyridoxine caused by microorganisms within tobacco phyllosphere.
Plant physiology and biochemistry : PPB (2014-11-15)
ShuoHao Huang, JianYun Zhang, Zhen Tao, Liang Lei, YongHui Yu, LongQuan Huang
ABSTRACT
Vitamin B6 (VB6) comprises six interconvertible pyridine compounds (vitamers), among which pyridoxal 5'-phosphate (PLP) is a coenzyme involved in a high diversity of biochemical reactions. In plants, PLP is de novo synthesized, and pyridoxine (PN) is usually maintained as the predominant B6 vitamer. Although the conversion from pyridoxal (PL) to PN catalyzed by PL reductase in plants has been confirmed, the enzyme itself remains largely unknown. We previously found pre-incubation at 35 °C dramatically enhanced PL reductase activity in tobacco leaf homogenate. In this study, we demonstrated that the increase in the reductase activity was a consequence of phyllosphere microbial proliferation. VB6 was detected from tobacco phyllosphere, and PL level was the highest among three non-phosphorylated B6 vitamers. When the sterile tobacco rich in PL were kept in an open, warm and humid environment to promote microorganism proliferation, a significant change from PL to PN was observed. Our results suggest that there may be a plant-microbe interaction in the conversion from PL to PN within tobacco phyllosphere.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid