Merck
  • Predictive value of serum actin-free Gc-globulin for complications and outcome in acute liver failure.
Predictive value of serum actin-free Gc-globulin for complications and outcome in acute liver failure.
Journal of viral hepatitis (2014-04-30)
A Bagchi, S Kumar, P C Ray, B C Das, P K Gumma, P Kar
ABSTRACT
This prospective study was designed to evaluate whether early changes in actin-free Gc-globulin levels were associated with complications and outcomes and to identify factors associated with persistent low actin-free Gc-globulin levels in acute liver failure (ALF). Thirty-two consecutive ALF patients admitted from October 2011 to December 2012 were followed up until death or complete recovery. All had serum actin-free Gc-globulin estimation at admission and at day three or expiry. Logistic regression analysis was performed to identify independent predictors of mortality. A receiver operating characteristic curve analysis was also performed. Nonsurvivors had significantly lower median actin-free Gc-globulin levels than survivors (87.32 vs 180 mg/L; P < 0.001). A receiver operating characteristic curve analysis revealed an area under curve (AUC) of 0.771 and showed that serum actin-free Gc-globulin level of ≤124 mg/L would predict mortality with 92% sensitivity and 71.4% specificity. Patients with lower serum actin-free Gc-globulin levels and decreasing trend in serum actin-free Gc-globulin levels were found to have more mortality and developed more complications. Logistic regression analysis showed that serum actin-free Gc-globulin, total leucocyte count and serum creatinine at admission were independent predictors of mortality. Incorporating these variables, a score predicting mortality risk at admission was derived. The scoring system was compared to MELD score and King's College Criteria as individual predictor of mortality. Serum actin-free Gc-globulin level at presentation is predictive of outcome and can be used for risk stratification. Its persistent low-level predicts mortality and is correlated with various complications.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Dibutyl phthalate, 99%