Merck
  • Using sheath-liquid reagents for capillary electrophoresis-mass spectrometry: application to the analysis of phenolic plant extracts.
Using sheath-liquid reagents for capillary electrophoresis-mass spectrometry: application to the analysis of phenolic plant extracts.
Electrophoresis (2014-10-14)
Leila Maringer, Elena Ibáñez, Wolfgang Buchberger, Christian W Klampfl, Tim J Causon
ABSTRACT
The combination of CE and MS is now a widely used tool that can provide a combination of high resolution separations with detailed structural information. Recently, we highlighted the benefits of an approach to add further functionality to this well-established hyphenated technique, namely the possibility to perform chemical reactions within the sheath-liquid of the CE-MS interface . Apart from using hydrogen/deuterium exchange for online determination of numbers of exchangeable protons, the addition of DPPH• (2,2-diphenyl-1-picrylhydrazyl) to the sheath-liquid can be used as a fast screening tool for studying antioxidant characteristics of individual components. Such a CE-MS methodology allows rapid and information-rich analysis with minimal reagent and sample consumption to be performed. In the present work, we demonstrate the applicability of this approach for the characterization of phenolic plant extracts from the Labiatae family, namely Rosmarinus officinalis and Melissa officinalis. Using the described approach, a wide range of compounds (15 and 13 phenolic compounds, respectively) could be confidently identified using a combination of high resolution CE-MS separations with implementation of online deuterium exchange and DPPH• reactions. These compounds included polyphenols, phenolic acids, and triterpene acids. In conjunction with online MS/MS experiments, extensive structural information for aglyconic and glycosylated antioxidants present in the extracts could be obtained using simple experimental changes, which can be carried out prior to the purchasing of expensive chemical standards or the time-consuming preparative isolation of individual compounds.
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Rosmarinic acid, 96%
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Carnosic acid from Rosmarinus officinalis, ≥91%, powder
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Rosmarinic acid, European Pharmacopoeia (EP) Reference Standard
Rosmarinic acid, primary reference standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis