Merck
  • Quantification of reactive oxygen species generation by photoexcitation of PEGylated quantum dots.
Quantification of reactive oxygen species generation by photoexcitation of PEGylated quantum dots.
Small (Weinheim an der Bergstrasse, Germany) (2014-08-29)
Elnaz Yaghini, Katharina F Pirker, Christopher W M Kay, Alexander M Seifalian, Alexander J MacRobert
ABSTRACT
Photocatalytic generation of reactive oxygen species (ROS) from quantum dots (QDs) has been widely reported yet quantitative studies of ROS formation and their quantum yields are lacking. This study investigates the generation of ROS by water soluble PEGylated CdSe/ZnS QDs with red emission. PEGylation of QDs is commonly used to confer water solubility and minimise uptake by organs of the reticuloendothelial system; therefore studies of ROS formation are of biomedical relevance. Using non-photolytic visible wavelength excitation, the superoxide anion radical is shown to be the primary ROS species generated with a quantum efficiency of 0.35%. The yield can be significantly enhanced in the presence of the electron donor, nicotinamide adenine dinucleotide (NADH), as demonstrated by oxygen consumption measurements and electron paramagnetic resonance spectroscopy with in situ illumination. Direct production of singlet oxygen is not detectable from the QDs alone. A comparison is made with ROS generation by the same QDs complexed with a sulfonated phthalocyanine which can generate singlet oxygen via Förster resonance energy transfer between the QDs and the phthalocyanine.
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%