Merck
  • Evaluation of the full evaporation technique for quantitative analysis of high boiling compounds with high affinity for apolar matrices.
Evaluation of the full evaporation technique for quantitative analysis of high boiling compounds with high affinity for apolar matrices.
Journal of chromatography. A (2014-05-17)
Niels van Boxtel, Kris Wolfs, Ann van Schepdael, Erwin Adams
ABSTRACT
In order to reduce inaccuracies due to possible matrix effects in conventional static headspace-gas chromatography (sHS-GC), it is standard practice to match the composition of calibration standards towards the composition of the sample to be analysed by adding blank matrix. However, the latter is not always available and in that case the full evaporation technique (FET) could be a solution. With FET a small sample volume is introduced in a HS vial and compounds of interest are completely evaporated. Hence no equilibrium between the condensed phase and vapour phase exists. Without the existence of an equilibrium, matrix effects are less likely to occur. Another issue often encountered with sHS-sampling is that low vapour pressure compounds with a high affinity for the dilution medium show a limited sensitivity. FET has proven to be an appropriate solution to address this problem too. In this work, the applicability of FET for the quantitative analysis of high boiling compounds in different complex apolar matrices is examined. Data show that FET is an excellent tool to overcome matrix effects often encountered with conventional sHS analysis. The tested method shows excellent accuracy with recovery values around 100% as well as repeatability with RSD values around 1% for the quantification of high boiling compounds (bp>200°C) such as camphor, menthol, methyl salicylate and ethyl salicylate in various matrices. LOQ values were found to be around 0.3μg per vial. Following validation of the technique, several topical pharmaceutical formulations like ThermoCream(®), Reflexspray(®), Vicks Vaporub(®) and Radosalil(®) were examined. For the latter, a comparison has been made with a sHS-method described in literature.
Product Description
Supelco
Density Standard 749 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Dodecane, anhydrous, ≥99%
Supelco
Dodecane, analytical standard
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
1-Octanol, suitable for HPLC, ≥99%
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Supelco
1-Octanol, analytical standard
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC