Merck
  • Determination of amphetamine and methylphenidate in exhaled breath of patients undergoing attention-deficit/hyperactivity disorder treatment.
Determination of amphetamine and methylphenidate in exhaled breath of patients undergoing attention-deficit/hyperactivity disorder treatment.
Therapeutic drug monitoring (2014-01-24)
Olof Beck, Niclas Stephanson, Sören Sandqvist, Johan Franck
ABSTRACT
It has been discovered recently that exogenous substances are detectable in exhaled breath after intake. Exhaled breath therefore constitutes a new possible matrix in clinical pharmacology and toxicology. The present work was aimed at exploring this possibility further by a study on patients treated for attention-deficit/hyperactivity disorder with D-amphetamine and methylphenidate. Thirteen patients (age range: 32-61 years; 5 women) were included in the study, and breath and urine samples were collected at different times in the dose interval. Analyses of breath and urine samples were done with liquid chromatography-mass spectrometry methods. Urine was examined for amphetamine, methylphenidate, and its metabolite ritalinic acid. Among the 9 patients who received D-amphetamine medication in daily doses of 20-100 mg, amphetamine was detected in all subjects in amounts ranging from 1200 to 30,800 picogram per filter. Among 8 patients receiving methylphenidate medication in daily doses of 80-400 mg, it was detected and quantified in 7 of the cases in amounts ranging from 150 to 10,400 picogram per filter and ritalinic acid was detected and quantified in 3 of the cases ranging from 35 to 360 picogram per filter. In 1 case, methylphenidate was only detectable in breath and urine, whereas ritalinic acid was quantifiable in urine, which could indicate noncompliance, with the 4 hours of dose regimen prescribed. In a number of cases, the sampling was performed 24 hours after the last dose intake. Identification of amphetamine and methylphenidate was based on correct chromatographic retention time and correct product ion ratio with detection performed in selected reaction monitoring mode. The results confirm that amphetamine is present in exhaled breath after intake and demonstrate for the first time the presence of methylphenidate and ritalinic acid after its intake. This gives further support to the potential use of exhaled breath for detecting drug intake.
Sigma-Aldrich
Ritalinic acid, 99% (CP)
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard