Merck
  • Noncompetitive inhibition of proton-coupled folate transporter by myricetin.
Noncompetitive inhibition of proton-coupled folate transporter by myricetin.
Drug metabolism and pharmacokinetics (2014-02-05)
Mai Furumiya, Katsuhisa Inoue, Chihiro Nishijima, Takahiro Yamashiro, Erina Inaoka, Kinya Ohta, Yayoi Hayashi, Hiroaki Yuasa
ABSTRACT
Myricetin is a flavonoid that has recently been suggested to interfere with the intestinal folate transport system. The present study was conducted to examine that possibility, focusing on its inhibitory effect on proton-coupled folate transporter (PCFT) as the molecular entity of the transport system. The uptake transport of folate was first examined in the Caco-2 cell as an intestinal epithelial cell model, and its carrier-mediated component, of which the Michaelis constant (Km) was 0.407 µM, was found to be noncompetitively inhibited by myricetin with an inhibition constant (Ki) of 61 µM. Consistent with that, folate transport by human PCFT stably expressed in Madin-Darby canine kidney II (MDCKII) cells, of which the Km was 1.246 µM, was also noncompetitively inhibited by myricetin with a Ki of 130 µM. Thus, myricetin was suggested to inhibit intestinal folate transport by acting noncompetitively on PCFT, although the Km and Ki were similarly shifted to some extent to be smaller in Caco-2 cells. Finally, epigallocatechin-3-gallate was also suggested to act in a noncompetitive manner as an inhibitory flavonoid. Care may need to be taken, therefore, in the ingestion of myricetin and some flavonoids to maintain the absorption of folate and antifolate drugs.
Product Description
Methotrexate, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(−)-Epigallocatechin gallate, analytical standard
Supelco
Myricetin, analytical standard
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Epigallocatechin gallate, primary reference standard
Supelco
Naringin, analytical standard
Sigma-Aldrich
Naringin, ≥90% (HPLC), from citrus fruit
Sigma-Aldrich
Naringin, ≥95% (HPLC)
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Naringin, European Pharmacopoeia (EP) Reference Standard
SAFC
Methotrexate
Sigma-Aldrich
(±)-Naringenin, ≥95%
Sigma-Aldrich
Naringenin, natural (US), 98%
Supelco
(±)-Naringenin, analytical standard
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard