Merck
  • Catalytic hydrothermal upgrading of crude bio-oils produced from different thermo-chemical conversion routes of microalgae.
Catalytic hydrothermal upgrading of crude bio-oils produced from different thermo-chemical conversion routes of microalgae.
Bioresource technology (2015-03-25)
Peigao Duan, Bing Wang, Yuping Xu
ABSTRACT
This study presents experimental results that compare the use of hydrothermal liquefaction (HTL), alcoholysis (Al), pyrolysis (Py) and hydropyrolysis (HPy) for the production of bio-oil from a microalga (Chlorella pyrenoidosa) and the catalytic hydrothermal upgrading of crude bio-oils produced by these four conversion routes. The yields and compositions of bio-oil, solid residue, and gases were evaluated and compared. HTL resulted in a bio-oil that has a higher energy density and superior fuel properties, such as thermal and storage stabilities, compared with the other three conversion routes. The N in crude bio-oils produced from Py and HPy is more easily removed than that in the bio-oils produced from HTL and Al. The upgraded bio-oils contain reduced amounts of certain O-containing and N-containing compounds and significantly increased saturated hydrocarbon contents. All of the upgraded bio-oils have a larger fraction boiling below 350°C than their corresponding crude bio-oils.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 51143095, Potassium nitrate, traceable to primary standards (LGC)
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Supelco
Sodium nitrate, analytical standard
Sigma-Aldrich
Sodium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Potassium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrate, ReagentPlus®, ≥99.0%