Merck
  • Clostridium difficile in retail baskets, trolleys, conveyor belts, and plastic bags in Saudi Arabia.
Clostridium difficile in retail baskets, trolleys, conveyor belts, and plastic bags in Saudi Arabia.
Saudi medical journal (2014-10-16)
Mohammed A Alqumber
ABSTRACT
To determine Clostridium difficile (C. difficile) prevalence on retail surfaces and shoppers plastic bags. From 20 June to 10 August 2011, in a cross-sectional epidemiological study, 17 supermarkets from 2 cities, Albaha and Altaif, Saudi Arabia were sampled. A total of 800 samples, which comprised 200 samples per surveyed surface, were studied. These included baskets, trolleys, conveyer belts, and outgoing shoppers' plastic bags. Clostridium difficile strains were isolated. The isolates were characterized using ribotyping and  polymerase chain reaction for the detection of toxin A (tcdA), toxin B (tcdB), binary toxin (cdtB), and toxin C (tcdC) genes. Susceptibility to antibiotics was determined on a Muller-Hinton agar with 5% sheep blood agar using E-tests. Overall, the C. difficile prevalence on sampled surfaces was 0.75%. The highest prevalence was found on retail baskets and trolleys, followed by plastic bags. A total of 5 different ribotypes were identified. Alterations in tcdC were detected in ribotype 027 and BT1. All the identified isolates were susceptible to vancomycin, but resistant to levofloxacin. In this study, C. difficile was present at a rate of 0.75% on supermarket surfaces. Spore disinfection of implicated surfaces may be necessary to control any community-acquired infections caused by this pathogen.
Product Description
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Sodium phosphate dibasic, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Supelco
Levofloxacin, analytical standard
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)