Merck
  • Uncovering a glycosyltransferase provides insights into the glycosylation step during macrolactin and bacillaene biosynthesis.
Uncovering a glycosyltransferase provides insights into the glycosylation step during macrolactin and bacillaene biosynthesis.
Chembiochem : a European journal of chemical biology (2014-10-30)
Wen Qin, Yang Liu, Pengfei Ren, Jun Zhang, Huayue Li, Li Tian, Wenli Li
ABSTRACT
Macrolactins (MLNs) have unique structural patterns containing a 24-membered ring lactone and diverse bioactivities. The MLN skeleton is biosynthesized via a trans-acyl transferase (AT) type I polyketide synthase (PKS) pathway, but the tailoring steps are still unknown. Herein, we report the identification of a glycosyltransferase (GT) gene bmmGT1, which is located at different locus from the MLN gene cluster in the genome of marine-derived Bacillus marinus B-9987, and its functional characterization as an MLN GT, thus affording five novel MLNs analogues. Surprisingly, this GT is also capable of catalyzing the glycosylation of bacillaenes (BAEs), which are the prototypes of trans-AT polyketides, thus suggesting broad substrate flexibility. These results provide the first significant insights into the glycosylation step in MLN and BAE biosynthetic pathways.
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Imidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration), free-flowing, Redi-Dri
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, FCC, FG
Supelco
Glycerol, analytical standard