Merck
  • An acellular cerebellar biological scaffold: Preparation, characterization, biocompatibility and effects on neural stem cells.
An acellular cerebellar biological scaffold: Preparation, characterization, biocompatibility and effects on neural stem cells.
Brain research bulletin (2015-03-21)
Tongming Zhu, Qisheng Tang, Yiwen Shen, Hailiang Tang, Luping Chen, Jianhong Zhu
ABSTRACT
Biomaterial and regenerative medical research has diversified and developed rapidly. A biological scaffold consisting of an extracellular matrix (ECM) functions not only as a supportive material but also as a regulator of cellular functions. Although decellularized scaffolds have been widely applied for the repair of non-central nervous system (CNS) tissues, their efficacy in the CNS has not been extensively investigated. In this report, we describe a dynamic decellularization protocol that combined intracardial perfusion and a series of treatments to effectively remove the cellular components from the cerebellum, which is a unique and relatively simple CNS structure. The resulting cerebellar scaffold retained neurosupportive proteins and growth factors and, when tested with neural stem cells (NSCs) in vitro, was found to be cytocompatible and to stimulate the proliferation and migration of these cells. NSCs that were cultured in vitro on the scaffold differentiated into neurons and astrocytes, as indicated by their expression of βIII-tubulin and glial fibrillary acidic protein (GFAP). Through subcutaneous and intracranial implantation experiments, this preliminary study demonstrated the in vivo biocompatibility of the cerebellar scaffold and indicated its potential for future applications. Thus, our study demonstrated that the cerebellar ECM scaffold provided tissue-specific advantages for regenerative medical applications.
Sigma-Aldrich
FGF-2 human, recombinant, expressed in insect cells, ≥85% (SDS-PAGE)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
EGF from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
EGF from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material