Merck
  • Uniaxial repetitive mechanical overloading induces influx of extracellular calcium and cytoskeleton disruption in human tenocytes.
Uniaxial repetitive mechanical overloading induces influx of extracellular calcium and cytoskeleton disruption in human tenocytes.
Cell and tissue research (2014-11-02)
Wan Chen, Yinshuan Deng, Jiqiang Zhang, Kanglai Tang
ABSTRACT
Tendon calcification is common in the Achilles tendon, and injuries affect not only athletes, but also the general population. However, the underlying cellular mechanisms are not yet fully understood. In this study, we isolated healthy human tenocytes and subjected them to uniaxial mechanical stretching (at 1.0 Hz) for various stretch times (4 h, 8 h, 12 h) or magnitudes (0%, 4%, 8%, 12%). The extracellular calcium chelator EGTA, calcium channel inhibitor MnCl2, nifedipine, or various doses of exogenous calcium were administered to these cells with or without mechanical overloading. The intracellular calcium concentration was determined by using a Fluo-3/AM fluorescence probe, and the cytoskeleton was revealed by F-actin Phalloidin staining. The intracellular calcium concentration increased in a magnitude- and time-dependent manner following stretching. These increases were suppressed by EGTA, MnCl2, or nifedipine. Additionally, cytoskeleton F-actin was disrupted significantly by stretching in a time-dependent manner. When extracellular calcium was applied, the intracellular calcium concentration increased, and F-actin was disrupted dramatically under mechanical stretching compared with non-stretched cells. Thus, repetitive mechanical overloading induces the accumulation of abnormally high concentrations of intracellular calcium resulting from extracellular calcium influx mediated, at least in part, by membrane calcium channels and finally causes cytoskeleton disorganization and tenocyte dysfunction. These findings provide novel experimental evidence for the pathology of tendon calcification and indicate that the blockade of calcium influx is a potential target for the prevention and treatment of calcific tendinopathy.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Fluorescein, free acid
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Supelco
Malachite Green oxalate salt, VETRANAL®, analytical standard