Merck
  • N-3 fatty acids improve body composition and insulin sensitivity during energy restriction in the rat.
N-3 fatty acids improve body composition and insulin sensitivity during energy restriction in the rat.
Prostaglandins, leukotrienes, and essential fatty acids (2014-08-31)
N Guelzim, J-F Huneau, V Mathé, S Tesseraud, J Mourot, N Simon, D Hermier
ABSTRACT
The hypothesis that n-3 polyunsaturated fatty acids (PUFA) could contribute to maintain muscle mass during energy restriction aiming to weight loss was tested in the rat, with special attention paid to insulin signalling. After 10 weeks on a diet rich in lipids and sucrose, male rats were energy restricted and fed diets rich in 18:1 n-9 (OLE), 18:3 n-3 (ALA) or n-3 long-chain (LC, >18 carbons) PUFA. After 4 weeks, they were killed after an insulin injection. Red blood cells, liver, and Gastrocnemius muscle were enriched in ALA in the ALA group, and in LC-PUFA in the ALA and LC groups. The LC diet resulted in a higher weight loss, without negative impact on the muscle weight. In parallel, hepatic phosphorylation of insulin receptor and IRS1 was the highest in this group. This suggests that the trend we observed in the preservation of protein homeostasis in the LC group is mediated, at least partly, by an enhancement of the early steps of insulin signalling resulting from cell membrane enrichment in n-3 PUFA.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Sodium fluoride, powder, 99.99% trace metals basis
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Supelco
Urea, analytical standard
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)