Merck
  • Intracisternal cyclodextrin prevents cerebellar dysfunction and Purkinje cell death in feline Niemann-Pick type C1 disease.
Intracisternal cyclodextrin prevents cerebellar dysfunction and Purkinje cell death in feline Niemann-Pick type C1 disease.
Science translational medicine (2015-02-27)
Charles H Vite, Jessica H Bagel, Gary P Swain, Maria Prociuk, Tracey U Sikora, Veronika M Stein, Patricia O'Donnell, Therese Ruane, Sarah Ward, Alexandra Crooks, Su Li, Elizabeth Mauldin, Susan Stellar, Marc De Meulder, Mark L Kao, Daniel S Ory, Cristin Davidson, Marie T Vanier, Steven U Walkley
ABSTRACT
Niemann-Pick type C1 (NPC) disease is a lysosomal storage disease caused by mutations in the NPC1 gene, leading to an increase in unesterified cholesterol and several sphingolipids, and resulting in hepatic disease and progressive neurological disease. We show that subcutaneous administration of the pharmaceutical excipient 2-hydroxypropyl-β-cyclodextrin (HPβCD) to cats with NPC disease ameliorated hepatic disease, but doses sufficient to reduce neurological disease resulted in pulmonary toxicity. However, direct administration of HPβCD into the cisterna magna of presymptomatic cats with NPC disease prevented the onset of cerebellar dysfunction for greater than a year and resulted in a reduction in Purkinje cell loss and near-normal concentrations of cholesterol and sphingolipids. Moreover, administration of intracisternal HPβCD to NPC cats with ongoing cerebellar dysfunction slowed disease progression, increased survival time, and decreased the accumulation of brain gangliosides. An increase in hearing threshold was identified as a potential adverse effect. These studies in a feline animal model have provided critical data on efficacy and safety of drug administration directly into the central nervous system that will be important for advancing HPβCD into clinical trials.
Product Description
Propofol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Propofol for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, powder
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
2,6-Diisopropylphenol, 97%