Merck
  • Protective effects of alpha lipoic acid versus N-acetylcysteine on ifosfamide-induced nephrotoxicity.
Protective effects of alpha lipoic acid versus N-acetylcysteine on ifosfamide-induced nephrotoxicity.
Toxicology and industrial health (2013-01-10)
Alaa El-Din E El-Sisi, Magda E El-Syaad, Karima I El-Desoky, Ethar A Moussa
ABSTRACT
Ifosfamide (IFO) is a highly effective chemotherapeutic agent for treating a variety of pediatric solid tumors. However, its use is limited due to its serious side effect on kidneys. The side-chain oxidation of IFO in renal tubular cells produces a reactive toxic metabolite that is believed to be responsible for its nephrotoxic effect. Therefore, this study was carried out to investigate the possible underlying mechanisms that may be involved in IFO-induced nephrotoxicity, including free radical generation and the possible role of alpha lipoic acid (ALA) versus N-acetylcysteine (NAC) in protection against this toxicity. Male albino rats were injected intraperitoneally with saline, IFO (50 mg/kg daily for 5 days), IFO + ALA (100 mg/kg daily for 8 days) and IFO + NAC (200 mg/kg daily for 8 days). Kidney malondialdehyde, nitric oxide and glutathione contents and serum biochemical parameters and histopathological analysis were determined. Both ALA and NAC markedly reduced the severity of renal dysfunction induced by IFO. NAC was more nephroprotective than ALA. This study suggests that oxidative stress is possibly involved in the IFO-induced nephrotoxicity in rats. The study also suggests the potential therapeutic role for ALA and NAC against IFO-induced nephrotoxicity.
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, ≥97.0% (AT)
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, BioReagent, suitable for cell culture, powder, ≥98%
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, ≥98%
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Supelco
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS reagent, >98%
Supelco
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS reagent, ≥98%
Sigma-Aldrich
N-(1-Naphthyl)ethylenediamine dihydrochloride, ≥98%
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanadium, powder, -100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%