Merck
  • Rapidly prototyped multi-scale electrodes to minimize the voltage requirements for bacterial cell lysis.
Rapidly prototyped multi-scale electrodes to minimize the voltage requirements for bacterial cell lysis.
The Analyst (2015-01-20)
Christine M Gabardo, Aaron M Kwong, Leyla Soleymani
ABSTRACT
Lab-on-a-chip systems used for nucleic acid based detection of bacteria rely on bacterial lysis for the release of cellular material. Although electrical lysis devices can be miniaturized for on-chip integration and reagent-free lysis, they often suffer from high voltage requirements, and rely on the use of off-chip voltage supplies. To overcome this barrier, we developed a rapid prototyping method for creating multi-scale electrodes that are structurally tuned for lowering the voltage needed for electrical bacterial lysis. These three-dimensional multi-scale electrodes – with micron scale reaction areas and nanoscale features – are fabricated using benchtop methods including craft cutting, polymer-induced wrinkling, and electrodeposition, which enable a lysis device to be designed, fabricated, and optimized in a matter of hours. These tunable electrodes show superior behaviour compared to lithographically-prepared electrodes in terms of lysis efficiency and voltage requirement. Successful extraction of nucleic acids from bacterial samples processed by these electrodes demonstrates the potential for these rapidly prototyped devices to be integrated within practical lab-on-a-chip systems.
Sigma-Aldrich
Propidium iodide solution
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether