Merck
  • Graphene oxide nanosheet wrapped white-emissive conjugated polymer nanoparticles.
Graphene oxide nanosheet wrapped white-emissive conjugated polymer nanoparticles.
ACS nano (2014-04-17)
Dong Youn Yoo, Nguyen Dien Kha Tu, Su Jin Lee, Eunji Lee, Seong-Ran Jeon, Sunyong Hwang, Ho Sun Lim, Jong Kyu Kim, Byeong Kwon Ju, Heesuk Kim, Jung Ah Lim
ABSTRACT
We have demonstrated the preparation of white-emissive conjugated polymer nanoparticles wrapped with graphene oxide (GO) nanosheets. Highly stable, GO-wrapped, poly(9,9-di-n-octylfluorenyl-2,7-diyl) nanoparticles (GO-PFO NPs) with diameters in the range 30-150 nm were successfully obtained by utilizing the GO nanosheets as an interface stabilizer in an emulsification process. The synthesized GO-PFO NPs exhibited unique white-emitting photoluminescence with a characteristic green-emissive broad band above 500 nm, which was distinct from the photoluminescent behavior of PFO NPs without GO. This green emission was deduced to originate from the presence of the GO nanosheet shell surrounding the PFO NPs, rather than from luminescence of GO itself or formation of keto defects in the PFO chain. PL decay analysis showed that the GO-wrapped PFO NPs had a longer luminescence lifetime in comparison to PFO NPs without GO, and highly efficient energy transfer to lower energy state induced by the GO occurred.
Product Description
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Hydrazine solution, 1.0 M in THF
Sigma-Aldrich
Hydrazine solution, 1 M in acetonitrile
Sigma-Aldrich
Hydrazine solution, 35 wt. % in H2O
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Hydrazine solution, 1.0 M in ethanol